Learn More
Cayman Chemical 1- 2 6-Xylyl-2-thIourea 50mg
SDP
Supplier:  Cayman Chemical 3937350 MG
Encompass_Preferred
An analytical reference standard categorized as an anesthetic metabolite; a metabolite of xylazine; has been used as a marker for xylazine doping in horses; intended for research and forensic applications
Catalog No. 50-291-7238