Learn More
Medchemexpress LLC 1,2-Benzisothiazole, 3-(1-piperazinyl)- | 87691-87-0 | 99.96% | 219.31 | 10MM/1ML
SDP
Supplier:  Medchemexpress LLC HYW00975410MM/1ML
Encompass_Preferred
Antidepressant agent 10 (Compound 4) is an orally active compound with IC50 values of 190 nM for 5-HT1A and 110 nM for 5-HT2. It can be used in the research of mental disorders.
  • Orally active compound
  • Can be used in the research of mental disorders
Catalog No. 50-004-51595