Learn More
Medchemexpress LLC 1,2-Benzisothiazole, 3-(1-piperazinyl)- | 87691-87-0 | 99.96% | 219.31 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00975450G
Encompass_Preferred
Antidepressant agent 10 (Compound 4) is an orally active compound primarily used in the research of mental disorders. It exhibits selective activity towards serotonin receptors, making it a valuable tool for studying neurological diseases. This agent is provided as an off-white to light yellow solid.
  • Orally active compound
  • IC50 values of 190 nM for 5-HT1A and 110 nM for 5-HT2
  • Utilized in research of mental disorders
  • Off-white to light yellow solid appearance
  • Purity of 99.96% via HPLC
Catalog No. 50-004-53346