Learn More
Strem, An Ascensus Company (S,S)-Ph-BPE min. 98% 100mg
SDP
Supplier:  Strem, An Ascensus Company 150474100MG
Encompass
CAS# 824395-67-7. 100mg. (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, min. 98% (S,S)-Ph-BPE. MFCD07369028. Molecular Weight: 506.60. Molecular Formula: (C16H16)PCH2CH2P(C16H16). Color/form: white solid Strem# 15-0474. http://www.strem.com/catalog/v/15-0474/
Catalog No. 50-901-09906