Learn More
Medchemexpress LLC (+)-1,2-bis((2S,5S)-2,5-diphenylphospholano)ethane | 824395-67-7 | MFCD07369028 | 98.0% | 506.60 | C34H36P2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW021016
Encompass_Preferred
(+)-1,2-bis((2S,5S)-2,5-diphenylphospholano)ethane is a chiral bisphospholane diphosphine ligand used in asymmetric catalysis and coordination chemistry. It appears as a white to off-white solid and is supplied with high chemical purity for research applications requiring stereoselective metal coordination.
  • Provides strong, stereoselective coordination to transition metals.
  • Used in asymmetric hydrogenation and other enantioselective transformations.
  • High chemical purity (98.0%) supports reproducible catalytic performance.
  • Molecular weight 506.60 and formula C34H36P2 for stoichiometric calculations.
  • Stable as a solid when stored under inert atmosphere at 4°C; in solution store at -80°C for long-term.
Catalog No. 50-003-02973