Learn More
Medchemexpress LLC (+)-1,2-bis((2S,5S)-2,5-diphenylphospholano)ethane | 824395-67-7 | MFCD07369028 | 98.0% | 506.60 g/mol | C34H36P2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW02101650MG
Encompass_Preferred
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane is a chiral phosphine ligand used in asymmetric catalysis, commonly applied in enantioselective hydrogenation and other stereoselective transformations. Supplied as a research-grade reagent with documented purity and handling information, it is intended for laboratory-scale synthesis and screening where high enantioselectivity is required.
  • High purity (98.0%) suitable for research applications.
  • Chiral (S,S) stereochemistry for enantioselective catalysis.
  • Stable when stored under nitrogen at 4°C.
  • In solvent stability: -80°C for 6 months, -20°C for 1 month (stored under nitrogen).
  • Available in small-scale quantities appropriate for reaction screening (25-100 mg).
Catalog No. 50-002-99508