Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Bis(3-(trifluoromethyl)phenyl)diselane | 53973-75-4 | 99.9% | C14H8F6Se2 | 10 MM * 1 ML in DMSO
SDP

Supplier:  Medchemexpress LLC HY4730810MM/1ML

Encompass_Preferred

1,2-Bis(3-(trifluoromethyl)phenyl)diselane is an active compound, intended for research use only, with a molecular weight of 448.12. It appears as a light yellow to yellow liquid. Soluble in DMSO at 100 mg/mL (223.15 mM), it may require ultrasonic treatment; newly opened, non-hygroscopic DMSO is recommended.

  • Intended for research use only
  • Not sold to patients
  • Molecular weight: 448.12
  • Appearance: Light yellow to yellow liquid
  • Shipping: Room temperature (continental US)
  • Storage: -20°C under nitrogen
  • Storage in solvent: -80°C for 6 months, or -20°C for 1 month (under nitrogen)
  • Solubility: 100 mg/mL (223.15 mM) in DMSO
  • Ultrasonic treatment may be needed for solubility
  • Use newly opened DMSO for optimal solubility
  • Aliquoting and storing stock solutions prevents freeze-thaw inactivation

Catalog No. 50-003-59297


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.