Learn More
Strem, An Ascensus Company CAS# 1663-45-2. 100g. 1,2-Bis(diphenylphosphino)ethane, 99% DIPHOS. MFCD00003047
SDP
Supplier:  Strem, An Ascensus Company 150200100G
Encompass
CAS# 1663-45-2. 100g. 1,2-Bis(diphenylphosphino)ethane, 99% DIPHOS. MFCD00003047. Molecular Weight: 398.43. Molecular Formula: (C6H5)2PCH2CH2P(C6H5)2. Color/form: white pwdr. Strem# 15-0200. http://www.strem.com/catalog/v/15-0200/
Catalog No. 50-901-09832