Learn More
Medchemexpress LLC 1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid acetoxymethyl ester | 126150-97-8 | MFCD00036696 | >95.0% | 764.68 g/mol | C34H40N2O18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10054550MG
Encompass_Preferred
BAPTA-AM is a membrane-permeable intracellular calcium chelator commonly used to buffer cytosolic Ca2+ in cell-based assays. It crosses cell membranes as an acetoxymethyl ester and is converted intracellularly to the active chelating form, reducing free Ca2+ concentration. The compound has reported inhibitory activity against hERG and other voltage-gated potassium channels in the low micromolar range and is supplied for research use with recommended low-temperature storage for stability.
  • Membrane-permeable intracellular Ca2+ chelator.
  • Converted intracellularly to an active Ca2+ chelator.
  • Buffers cytosolic calcium in cell-based assays.
  • Exhibits micromolar inhibition of certain K+ channels.
  • Supplied as a powder with recommended low-temperature storage.
Catalog No. 50-002-12760