Learn More
Medchemexpress LLC 1-(2-Chlorophenyl)-2-((2-(methyl-d3)propan-2-yl-1,1,1,3,3,3-d6)amino)ethan-1-ol hydrochloride | 1325559-14-5 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1810S1MG
Encompass_Preferred
Tulobuterol-d9 (hydrochloride) is the deuterium labeled Tulobuterol, a long-acting β2-adrenoceptor agonist. It is known to reduce the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma. This compound also functions as a sympathomimetic agent when used as a transdermal patch, enhancing normal diaphragm muscle strength. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11913