Learn More
Medchemexpress LLC 1 2-Cyclohexylenedin 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HYW10271410MM/1ML
Encompass_Preferred
1 2-Cyclohexylenedinitrilotetraacetic acid (CDTA) is a chelating agent 1 2-Cyclohexylenedinitrilotetraacetic acid has an ability to remove manganese from brain and liver (in vivo) and their sub-cellular fractions (in vitro) of rats pretreated with manganese sulphate[1]
Catalog No. 50-002-06511