Learn More
Medchemexpress LLC 1 2-Cyclohexylenedin 500mg
SDP
Supplier:  Medchemexpress LLC HYW102714500MG
Encompass_Preferred
1 2-Cyclohexylenedinitrilotetraacetic acid (CDTA) is a chelating agent 1 2-Cyclohexylenedinitrilotetraacetic acid has an ability to remove manganese from brain and liver (in vivo) and their sub-cellular fractions (in vitro) of rats pretreated with manganese sulphate[1]
Catalog No. 50-002-05601