Learn More
Medchemexpress LLC 1,2-Diaminoanthraquinone | 1758-68-5 | 91.1% | 238.25 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW013435
Encompass_Preferred
1,2-Diaminoanthraquinone is a sensitive, specific, and nontoxic nitric oxide (NO) fluorescent probe. It can be used to detect NO productions in live cells and animals. Its maximum absorption is at about 540 nm and it has a detection limit of 5 μM for NO.
  • Sensitive, specific, and nontoxic nitric oxide (NO) fluorescent probe
  • Can be used to detect NO productions in live cells and animals
  • Maximum absorption at about 540 nm
  • Detection limit of 5 μM for NO
  • Spectrally resolved using fluorescence spectroscopy and confocal microscopy
  • Reacts with NO in the presence of oxygen in the intracellular environment to yield DAA-TZ
Catalog No. 50-004-51310