Learn More
Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00037499 | 99.0% | 191.89 g/mol | BrCD2CD2Br | 5 G
SDP
Supplier:  Medchemexpress LLC HYW578702S5G
Encompass_Preferred
1,2-Dibromoethane-d4 is the deuterated isotopologue of 1,2-dibromoethane in which four hydrogen atoms are replaced by deuterium. It is supplied as a small-quantity reagent for analytical and synthetic applications where isotopic labeling is required, such as NMR solvents, internal standards, tracer studies, and mechanistic investigations.
  • Deuterium-labeled (d4) isotopologue for isotopic tracing.
  • Suitable as an internal standard for NMR and mass spectrometry.
  • Useful in mechanistic and synthetic studies requiring isotopic enrichment.
  • Provided in small laboratory-scale packaging for research use.
  • Chemical formula BrCD2CD2Br and CAS number 22581-63-1 for unambiguous identification.
Catalog No. 50-002-84454