Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dichloro-3-methylbenzene-d3 | 1219798-57-8 | 164.05 | C7H3D3Cl2 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW015427S

Encompass_Preferred

1,2-Dichloro-3-methylbenzene-d3 is the deuterium-labeled analogue of 1,2-dichloro-3-methylbenzene, intended for use as an isotope-labeled standard or tracer in analytical workflows. It provides a stable heavy-isotope tag for quantitative LC-MS, metabolic tracing, and pharmacokinetic studies.

  • Deuterium-labeled (d3) compound for isotope tracing and internal standard use.
  • Molecular formula C7H3D3Cl2 and molecular weight 164.05.
  • Suitable for LC-MS quantitation and metabolic profiling.
  • Storage: pure form -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Useful for tracer studies and improving assay accuracy.

Catalog No. 50-003-03422


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.