Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC Reverse abasic amidite | 5 g | CAS 401813-16-9 | MDL MFCD29904575

Supplier:  STA PHARMACEUTICAL US LLC STAAM105555GM

Encompass

Reverse abasic amidite is a Amidite reagent (Subcategory: Abasic Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 401813-16-9
- MDL: MFCD29904575
- InChIKey: WPNQXLQBMSUHFX-JUWSMZLESA-N
- Molecular Weight: 620.726761998
- Molecular Formula: C35H45N2O6P
- Purity: ≥98%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: ((2R,3S)-3-(bis(4-methoxyphenyl)(phenyl)methoxy)tetrahydrofuran-2-yl)methyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OC[C@H]1OCC[C@@H]1OC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)OCCC#N)C(C)C)C

Catalog No. 50-236-2932


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.