Learn More
Medchemexpress LLC 1,2-diheptanoyl-sn-glycero-3-phosphocholine | 39036-04-9 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12205650MG
Encompass_Preferred
1,2-diheptanoyl-sn-glycero-3-phosphocholine is a synthetic short-chain phosphatidylcholine used as a lipid detergent and model membrane component in biochemical and biophysical research. It has defined molecular properties and is supplied at high purity for laboratory applications.
  • Short-chain phosphatidylcholine suitable for micelle formation and membrane mimetics.
  • High purity for analytical and biophysical assays.
  • Defined molecular weight and formula for accurate stoichiometry.
  • Available in multiple small pack sizes for lab-scale experiments.
  • Compatible with common organic solvents for lipid dispersion preparation.
Catalog No. 50-002-31574