Learn More
Medchemexpress LLC 1,2-dimethoxybenzene-d2 | 203645-56-1 | 99.7% | 140.18 g/mol | C8H8D2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1812S110MG
Encompass_Preferred
Veratrole-d2 (1,2-dimethoxybenzene-d2) is a deuterium-labeled isotope standard used in analytical chemistry and metabolic studies. It serves as an internal standard for mass spectrometry and gas chromatography and is applied in research on cytochrome P450-mediated metabolism and plant volatile analysis.
  • Deuterium-labeled internal standard for MS and GC analyses.
  • High isotopic purity for accurate quantitation and tracing.
  • Supplied as a small, stable solid for convenient handling and storage.
  • Applicable to metabolic studies, tracer experiments, and volatile organic compound research.
Catalog No. 50-002-97247