Learn More
Medchemexpress LLC 1,2-dimethoxybenzene-d6 | 24658-24-0 | 98.7% | 144.20 g/mol | C8H4D6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1812S310MG
Encompass_Preferred
Veratrole-d6 is the deuterium-labeled form of veratrole (1,2-dimethoxybenzene) supplied as an analytical isotope standard for use in mass spectrometry, metabolite studies, and cytochrome P450 research. It is provided with high isotopic and chemical purity for reliable analytical performance.
  • Deuterium-labeled internal standard for analytical assays
  • Molecular formula C8H4D6O2 and molecular weight 144.20 g/mol
  • High chemical purity (98.7%) for reproducible results
  • Suitable for metabolite profiling and cytochrome P450 studies
  • Available in small milligram packages for analytical use
Catalog No. 50-002-97825