Learn More
Medchemexpress LLC 1,2-dimyristoyl-rac-glycero-3-phosphocholine | 18656-38-7 | MFCD00066562 | ≥98.0% | 677.93 | C36H72NO8P | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW14259625MG
Encompass_Preferred
1,2-Dimyristoyl-rac-glycero-3-phosphocholine is a zwitterionic phospholipid commonly used as a simple model of eukaryotic cell membranes in biophysical studies and liposome preparation. It provides a neutral-charged bilayer environment for membrane protein and lipid interaction experiments. Typical specifications include high purity (≥98.0%), molecular weight 677.93 g/mol, and formula C36H72NO8P.
  • Suitable as a membrane mimic for biophysical experiments.
  • Supports liposome and vesicle formation.
  • Neutral headgroup mimics eukaryotic plasma membranes.
  • High purity for reproducible experimental results.
  • Compatible with standard lipid handling and solvent systems.
Catalog No. 50-002-83779