Learn More
Medchemexpress LLC 1,2-dimyristoyl-rac-glycero-3-phosphocholine | 18656-38-7 | MFCD00066562 | ≥98.0% | 677.93 g/mol | C36H72NO8P | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW142596250MG
Encompass_Preferred
1,2-Dimyristoyl-rac-glycero-3-phosphocholine (DMPC) is a zwitterionic phospholipid used as a simple model of eukaryotic plasma membranes. It readily forms bilayers and liposomes with a neutral surface charge, making it suitable for membrane biophysics, liposome formulation, and studies of lipid-protein interactions where a reproducible, neutral phospholipid matrix is required.
  • Forms stable bilayers and liposomes for membrane models.
  • Mimics the neutral charge of eukaryotic plasma membranes.
  • Suitable for membrane biophysics and lipid-protein interaction studies.
  • High purity for reproducible experimental results.
  • Available in multiple sizes for small- and large-scale experiments.
Catalog No. 50-002-83028