Learn More
Medchemexpress LLC 1,2-dimyristoyl-rac-glycero-3-phosphocholine | 18656-38-7 | MFCD00066562 | ≥98.0% | 677.93 g·mol⁻¹ | C36H72NO8P | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW14259650MG
Encompass_Preferred
1,2-dimyristoyl-rac-glycero-3-phosphocholine (DMPC) is a zwitterionic phospholipid commonly used as a simple eukaryotic membrane mimic for lipid bilayer and liposome research. It is used in membrane biophysics studies, liposome preparation, and membrane protein reconstitution, and is supplied at high purity suitable for analytical and experimental applications.
  • Zwitterionic phospholipid that mimics the neutral surface charge of eukaryotic plasma membranes.
  • High purity (≥98.0%) suitable for biochemical and biophysical assays.
  • Available in small-quantity packages for research-scale experiments.
  • Compatible with liposome formation and membrane protein reconstitution protocols.
  • Well-characterized molecular weight and formula for reproducible experimental design.
Catalog No. 50-002-82300