Learn More
Medchemexpress LLC 1,2-Dioleoyl-3-linoleoyl-rac-glycerol | 2190-20-7 | 5 MG
Supplier: Medchemexpress LLC HYN79535MG
1,2-Dioleoyl-3-linoleoyl-rac-glycerol (OOL) is a triacylglycerol that contains oleic acid at the sn-1 and sn-2 positions and linoleic acid at the sn-3 position. It is found in various seed and vegetable oils, such as pumpkin seed, olive, and sesame oils.
- Contains oleic acid at sn-1 and sn-2 positions
- Contains linoleic acid at sn-3 position
- Found in various seed and vegetable oils