Learn More
Medchemexpress LLC 1,2-Dioleoyl-sn-glycerol (sn-1,2-Dioleoylglycerol) | 24529-88-2 | 99.96% | C39H72O5 | 100 MG
Supplier: Medchemexpress LLC HY141572100MG
1,2-Dioleoyl-sn-glycerol (sn-1,2-Dioleoylglycerol) is a glycerolipid found on the plasma membrane. It is composed of two fatty acid chains linked to a single glycerol molecule via an ester bond. This compound serves as an analog of diacylglycerol (DAG), a second messenger that activates protein kinase C. It has also been utilized as a source of diacylglycerol in the diacylglycerol O-acyltransferase 1 (DGAT1) assay and can induce acrosome reaction in human sperm.
- Glycerolipid located on the plasma membrane.
- Analog of the protein kinase C-activating second messenger DAG.
- Used as a source of diacylglycerol in the diacylglycerol O-acyltransferase 1 (DGAT1) assay.
- Can induce acrosome reaction in human sperm.
- High purity of 99.96%.
- Supplied as a liquid with a density of 0.9218 g/cm³ and a colorless to light yellow appearance.