Learn More
Medchemexpress LLC 1,2-dipalmitoyl-sn-glycero-3-phosphoglycerol sodium | 200880-41-7 | MFCD00070068 | 98.0% | 744.95 g/mol | C38H74NaO10P | 1G
SDP
Supplier:  Medchemexpress LLC HY1259401G
Encompass_Preferred
DPPG (1,2-dipalmitoyl-sn-glycero-3-phosphoglycerol sodium) is an anionic phospholipid used to prepare liposomes, micelles, and artificial membranes for membrane biophysics and biochemical research. Supplied as the sodium salt in a white to off-white solid form, it is commonly used to model lipid bilayers and investigate lipid-protein interactions.
  • Supports formation of liposomes and bilayers for membrane modeling
  • Useful for studies of lipid-protein interactions
  • Anionic phospholipid with sodium counterion for improved solubility
  • High purity suitable for research applications
  • White to off-white solid compatible with common organic solvents
Catalog No. 50-002-35318