Learn More
Avanti Polar Lipids DSPE PEG(2000)-N-Cy5, 2315262-02-1, Chloroform, 1mg, 1mg/ml 1mL
Supplier:  Avanti Polar Lipids 810891C1MG
Encompass
Molecular Formula C164H299N4O55P (average MW due to polydispersity of PEG), Purity >99%, Formula Weight 3238.143, Storage Temperature -20°C, 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000]-N-(Cyanine 5)
Catalog No. NC1681749