Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphocholine-d9 | 326495-37-8 | 799.22 Da | C44H79D9NO8P | 1 MG

Supplier: Medchemexpress LLC HYW040193S51MG
1,2-distearoyl-sn-glycero-3-phosphocholine-d9 is a deuterium-labeled phosphatidylcholine lipid in which the choline headgroup contains nine deuterium atoms (d9). It is used as an isotopic tracer and labeled lipid in studies of lipid bilayers, liposomes, and lipid nanoparticles.
- Deuterated choline headgroup (d9) for isotopic tracing.
- Molecular formula: C44H79D9NO8P.
- Molecular weight: 799.22 Da.
- Soluble in chloroform; provided as a dry powder.
- Store dry at 4 °C; stock solutions should be frozen at -20 °C or below.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.