Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-5000] | 1236288-25-7 | 95.7% | 5,000 Da (average) | C63H104N9O15P | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW440890100MG
Encompass_Preferred
DSPE-PEG-Folate, MW 5000 is a folic acid-conjugated PEGylated phospholipid used to introduce folate targeting to liposomes, micelles, and other lipid-based formulations. The amphiphilic conjugate displays folate on particle surfaces for receptor-mediated uptake and provides PEG-based steric stabilization for improved circulation and colloidal stability.
  • Facilitates targeting to folate receptor-expressing cells
  • Incorporates into lipid bilayers and micelles
  • Adds PEG steric stabilization to nanoparticle surfaces
  • Suitable for receptor-mediated uptake and targeted delivery research
  • Available in multiple pack sizes for research use
Catalog No. 50-002-81151