Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine N-hydroxysuccinimide | 1383932-86-2 | 95.0% | 945.21 | C49H89N2O13P | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44101150MG
Encompass_Preferred
DSPE-NHS is an N-hydroxysuccinimide (NHS) ester-functionalized phospholipid for bioconjugation and liposome formulation. It self-assembles into lipid bilayers in aqueous solution, and the NHS ester reacts with primary amines on proteins or peptides to form stable amide bonds, enabling antibody labeling and nanoparticle surface modification.
  • Enables covalent coupling to primary amines via NHS ester chemistry.
  • Self-assembles into lipid bilayers for liposome formation.
  • Supports antibody labeling and nanoparticle surface functionalization.
  • Stable under recommended storage when kept dry and protected from light.
  • Supplied in laboratory-scale quantities suitable for research use.
Catalog No. 50-002-81716