Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-polyethylene glycol maleimide | 474922-22-0 | MFCD11501046 | >95.0% | 3,400 Da | (C2H4O)nC52H93N3NaO13P | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW441009A25MG
Encompass_Preferred
DSPE-PEG-Maleimide (MW 3400) ammonium is a phospholipid-PEG conjugate terminating in a maleimide reactive group (ammonium salt) used to functionalize lipid nanoparticle surfaces. It enables thiol-specific coupling of peptides, proteins, or small molecules to liposomes and nanostructured lipid carriers, providing steric stabilization and targeted-surface modification for drug delivery research.
  • Functional maleimide group for thiol-specific conjugation.
  • DSPE phospholipid anchor for stable insertion into lipid bilayers.
  • PEG chain with nominal molecular weight 3,400 Da for steric stabilization.
  • Ammonium salt form to aid solubility and handling.
  • Intended for liposome and nanostructured lipid carrier preparation.
  • Purity >95% (vendor specification).
  • Polymeric PEG represented as (C2H4O)n; composition varies with chain length.
Catalog No. 50-002-81139