Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine-d13 | 803.23 g/mol | C44H75D13NO8P | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW040193S61MG

Encompass_Preferred

DSPC-d13 (1,2-distearoyl-sn-glycero-3-phosphorylcholine-d13) is a deuterium-labeled phospholipid used as a tracer and analytical standard and as a lipid component in liposome and lipid nanoparticle formulations.

  • Deuterium labeled (D13), contains 13 deuterium atoms.
  • Molecular weight 803.23 g/mol.
  • Molecular formula C44H75D13NO8P.
  • Available as a 1 mg sample.
  • Typical applications include liposome and lipid nanoparticle formulation, tracer studies, and mass spectrometry internal standards.

Catalog No. 50-002-08698


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.