Learn More
Medchemexpress LLC 1,2-Ethanediol, 1-[4-hydroxy-3-(methoxy-d3)phenyl]- | 74495-72-0 | 99.5% | 187.21 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HY113323S2.5MG
Encompass_Preferred
3-Methoxy-4-hydroxyphenylglycol-d3 is the deuterium labeled 3-Methoxy-4-hydroxyphenylglycol.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-37858