Learn More
Medchemexpress LLC 1-(2-fluorophenyl)-3-(naphthalen-2-yl)prop-2-en-1-one | 867294-23-3 | 98.0% | 276.30 g/mol | C19H13FO | 10 MG

Supplier: Medchemexpress LLC HY4782510MG
WAY-660222 is a research-stage small-molecule bioactive compound supplied for laboratory use. It has a reported purity of 98.0%, molecular formula C19H13FO, and molecular weight 276.30 g/mol. The product is provided in milligram quantities with supporting documentation to assist experimental use, and should be stored per handling instructions to preserve stability.
- High reported purity suitable for research applications.
- Molecular formula C19H13FO; molecular weight 276.30 g/mol.
- Provided in milligram-scale quantities for assay and synthesis work.
- Documentation includes certificate of analysis, safety data sheet, and H NMR.
- Recommended storage: protect from light and follow supplier temperature guidance.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.