Learn More
Medchemexpress LLC 1-(2-Hydroxyethyl)piperazine | 103-76-4 | 99.5% | 130.19 | 100 G
SDP
Supplier:  Medchemexpress LLC HY41939100G
Encompass_Preferred
1-(2-Hydroxyethyl)piperazine is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. It is intended for research use only and not for sale to patients.
  • It is a biochemical reagent.
  • It can be used as a biological material.
  • It can be used as an organic compound for life science-related research.
  • 1-Piperazineethanol is a hydroxyalkyl substituted piperazine.
  • It is used in the preparation of pharmaceutical compounds.
  • It serves as an intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, and pharmaceuticals.
  • It is also used to synthesize triethylenediamine.
Catalog No. 50-004-04103