Learn More
Medchemexpress LLC 1-(2-Hydroxyethyl)piperazine | 103-76-4 | 99.5% | 130.19 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW012728
Encompass_Preferred
1-(2-Hydroxyethyl)piperazine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a hydroxyalkyl substituted piperazine used in the preparation of pharmaceutical compounds. It is also used as an intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, and pharmaceuticals. Additionally, it is used to synthesize triethylenediamine.
  • Biochemical reagent for life science research
  • Used in preparation of pharmaceutical compounds
  • Intermediate for polyurethane catalysts
  • Intermediate for corrosion inhibitors
  • Intermediate for surfactants
  • Intermediate for synthetic fibers
  • Intermediate for pharmaceuticals
  • Used to synthesize triethylenediamine
Catalog No. 50-003-66809