Learn More
Medchemexpress LLC 1-(2-Hydroxyethyl)piperidine | 3040-44-6 | ≥95.0% | 129.20 | 500 G

Supplier: Medchemexpress LLC HYW012728500G
2-(Piperidin-1-yl)ethanol is a biochemical reagent suitable for life science research. This organic compound is supplied as a liquid with a light yellow to brown color, offering a purity of ≥95.0% and a molecular weight of 129.20.
- Suitable for life science related research.
- Organic compound.
- Supplied as a liquid with a light yellow to brown color.
- Purity of ≥95.0%.
- Molecular weight of 129.20.
- Recommended storage at 4°C, sealed, away from moisture and light.
- For storage in solvent: -80°C for 6 months; -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.