Learn More
Medchemexpress LLC [1-[2-[(methylsulfonyl)amino]ethyl]-4-piperidinyl]methyl 1-methyl-1H-indole-3-carboxylate | 144625-51-4 | MFCD00912901 | 100.0% | 393.50 g/mol | C19H27N3O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10315210MM 1ML
Encompass_Preferred
GR 113808 is a potent and selective 5-HT4 (serotonin) receptor antagonist provided as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications. The product is supplied with analytical documentation to support use in in vitro assays.
  • Potent and selective 5-HT4 receptor antagonist (pKb ≈ 8.8).
  • Ready-to-use 10 mM solution in DMSO for in vitro applications.
  • High analytical purity suitable for research assays.
  • Provided with certificate of analysis and safety data sheet.
  • Compact 1 mL volume facilitates easy handling and storage.
Catalog No. 50-002-47488