Learn More
Medchemexpress LLC 1-(2-Pyridylazo)-2-naphthol | 85-85-8 | ≥90.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0223
Encompass_Preferred
1-(2-Pyridylazo)-2-naphthol is an azo compound used as a chelating precipitant, flocculant, auxiliary complexing agent, and as a ligand for anchoring on other supports to introduce chelating properties. It is for research use only.
  • Used as a chelating precipitant
  • Functions as a flocculant
  • Auxiliary complexing agent
  • Ligand for anchoring on other supports
  • Introduces chelating property
  • Suitable for research use
Catalog No. 50-004-14454