Learn More
Medchemexpress LLC 1,2-pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S,4R)- | 74844-91-0 | >=98.0% | 245.28 g/mol | C11H19NO5 | 500 G

Supplier: Medchemexpress LLC HY65039500G
N-Boc-4-hydroxy-L-proline methyl ester used as a linker building block for medicinal chemistry and bioconjugation. Supplied as a white to off-white solid with high purity suitable for synthesis and analytical work.
- used as a non-cleavable ADC linker and as a PROTAC linker building block.
- white to off-white solid physical form, suitable for handling and storage.
- purity ≥98.0% by NMR, supporting synthesis and analytical applications.
- molecular weight 245.28 g/mol and formula C11H19NO5 for formulation and calculation.
- available in large pack sizes for scale-up and bulk synthesis needs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.