Learn More
Cayman Chemical 1 24dIhydroxy VItamIn D3hy 5mg
SDP
Supplier:  Cayman Chemical 120795 MG
Encompass_Preferred
An analog of 1,24-dihydroxy vitamin D3 which, when used in a 4 μg/g ointment, suppresses psoriasis; inhibits the growth of both normal and psoriatic human keratinocytes (IC50 ~ 10 nM) via the vitamin D receptor; suppresses the production of RANTES and IL-8 by human dermal fibroblasts
Catalog No. 50-295-4422