Learn More
Cayman Chemical 1 2DIheptadecnoylsnglycer 50mg
SDP
Supplier:  Cayman Chemical 2463050 MG
Encompass_Preferred
A synthetic phosphatidylcholine; contains 17:0 fatty acids at the sn-1 and sn-2 positions; has been used as an internal standard for the quantification of phosphatidylcholines by LC-MS
Catalog No. 50-293-4029