Learn More
Cayman Chemical 1 2DImrIstol3EIcosapenta 10mg
SDP
Supplier:  Cayman Chemical 2682610 MG
Encompass_Preferred
A triacylglycerol; relative quantities in the alga T. minutus positively correlate with increases in temperature
Catalog No. 50-293-4201