Learn More
Cayman Chemical 1 2DImyrIstoyl3Lauroylrac 10mg
SDP
Supplier:  Cayman Chemical 2886110 MG
Encompass_Preferred
A triacylglycerol; has been found in butterfat, aged cheese, and date seed oil
Catalog No. 50-293-7025