Learn More
Cayman Chemical 1 2DImyrIstoyld54snglycer 10mg
SDP
Supplier:  Cayman Chemical 2875310 MG
Encompass_Preferred
An internal standard for the quantification of 1,2-dimyristoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-293-8745