Learn More
Cayman Chemical 1 2DImyrIstoylracglycerol 1g
SDP
Supplier:  Cayman Chemical 281711 G
Encompass_Preferred
A diacylglycerol; increases phosphorylation of the insulin receptor in IM-9 cells at 100 μg/ml; reduces tumor weight in an LY5178Y mouse xenograft model at 0.5 mg/kg
Catalog No. 50-293-9226