Learn More
Cayman Chemical 1 2DIoleoyl3dImethylamonI 10mg
SDP
Supplier:  Cayman Chemical 2572610 MG
Encompass_Preferred
An ionizable cationic lipid (pKa = 5.59 in a TNS binding assay); has been used as a component in liposomes that can be used to encapsulate siRNA, immunostimulatory oligodeoxynucleotides, antisense oligonucleotides, or chemotherapeutic agents for in vitro and in vivo delivery
Catalog No. 50-293-6050