Learn More
Cayman Chemical 1 2DIoleoyl3Stearoylracg 10mg
SDP
Supplier:  Cayman Chemical 2707110 MG
Encompass_Preferred
A triacylglycerol; has been found in sunflower, corn, and soybean oils, as well as ostrich oil
Catalog No. 50-293-8778