Learn More
Cayman Chemical 1 2DIoleoylsnglycero3PA 10mg
SDP
Supplier:  Cayman Chemical 3427410 MG
Encompass_Preferred
A phospholipid; has been used in the generation of micelles, liposomes, and other artificial membranes
Catalog No. 50-294-6534