Learn More
Cayman Chemical 1 2DIpalmItoyl13Csnglycer 10mg
SDP
Supplier:  Cayman Chemical 2913310 MG
Encompass_Preferred
An internal standard for the quantification of 1,2-dipalmitoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-293-9931