Learn More
Cayman Chemical 1 2DIpalmItoyl3ArachIdony 10mg
SDP
Supplier:  Cayman Chemical 2716110 MG
Encompass_Preferred
A triacylglycerol containing palmitic acid at the sn-1 and sn-2 positions and arachidonic acid at the sn-3 position
Catalog No. 50-293-9568